missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Tris(trifluoromethyl)benzene, TCI America™

Manufacturer:  TCI America T283025G

 View more versions of this product

Catalog No. T283025G



Specifications

Specifications

1,3,5-Tris(trifluoromethyl)benzene
Colorless
729-81-7
MFCD00054722
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
123085
Clear Liquid at 20°C
120°C
25g
C9H3F9
1993
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
282.109
282.11
≥98.0% (GC)

Chemical Identifiers

729-81-7
282.109
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
C9H3F9
MFCD00054722
123085
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents