missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Tris(4-iodophenyl)benzene 98.0+%, TCI America™
$220.65 - $764.79
Chemical Identifiers
CAS | 151417-38-8 |
---|---|
Molecular Formula | C24H15I3 |
Molecular Weight (g/mol) | 684.097 |
MDL Number | MFCD07369795 |
InChI Key | KGLWDSJGGFTHHD-UHFFFAOYSA-N |
PubChem CID | 10842293 |
IUPAC Name | 1,3,5-tris(4-iodophenyl)benzene |
SMILES | C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)I)C4=CC=C(C=C4)I)I |
Chemical Identifiers
151417-38-8 | |
684.097 | |
KGLWDSJGGFTHHD-UHFFFAOYSA-N | |
1,3,5-tris(4-iodophenyl)benzene |
C24H15I3 | |
MFCD07369795 | |
10842293 | |
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)I)C4=CC=C(C=C4)I)I |
Specifications
151417-38-8 | |
White-Yellow | |
MFCD07369795 | |
KGLWDSJGGFTHHD-UHFFFAOYSA-N | |
1,3,5-tris(4-iodophenyl)benzene | |
10842293 | |
≥98.0% (HPLC) | |
1,3,5-Tris(4-iodophenyl)benzene |
266°C | |
C24H15I3 | |
1 g | |
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)I)C4=CC=C(C=C4)I)I | |
684.097 | |
684.10 | |
Crystalline Powder |
Safety and Handling
TSCA : No