missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Tris(4-bromophenyl)benzene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3763603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,3,5-Tris(4-bromophenyl)benzene | |
261°C to 265°C | |
C24H15Br3 | |
1 g | |
HJQRITCAXSBOPC-UHFFFAOYSA-N | |
1,3,5-tris(4-bromophenyl)benzene | |
232761 | |
97% |
7511-49-1 | |
Brown to Yellow | |
MFCD00362911 | |
1,3,5-tris 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1':3',1-terphenyl, 1,1':3',1-terphenyl, 4,4-dibromo-5'-4-bromophenyl, 4'-bromo-3,5-bis 4-bromophenyl-1,1'-biphenyl, acmc-209ox3, ksc496e7d, 1,3,5-tris p-bromophenyl benzene, 1,3,5-tri 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1' | |
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br | |
543.096 | |
543.1 | |
Crystalline Powder |
Chemical Identifiers
7511-49-1 | |
543.096 | |
HJQRITCAXSBOPC-UHFFFAOYSA-N | |
232761 | |
C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br |
C24H15Br3 | |
MFCD00362911 | |
1,3,5-tris 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1':3',1-terphenyl, 1,1':3',1-terphenyl, 4,4-dibromo-5'-4-bromophenyl, 4'-bromo-3,5-bis 4-bromophenyl-1,1'-biphenyl, acmc-209ox3, ksc496e7d, 1,3,5-tris p-bromophenyl benzene, 1,3,5-tri 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1' | |
1,3,5-tris(4-bromophenyl)benzene |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only