missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Tris(3,5-dibromophenyl)benzene, TCI America™

Manufacturer:  TCI America T32131G

 View more versions of this product

Catalog No. T32131G



Specifications

Specifications

1,3,5-Tris(3,5-dibromophenyl)benzene
315°C
29102-67-8
MFCD13193235
C1=C(C=C(C=C1C2=CC(=CC(=C2)Br)Br)C3=CC(=CC(=C3)Br)Br)C4=CC(=CC(=C4)Br)Br
779.784
779.78
≥95.0% (HPLC)
Undesignated
1g
C24H12Br6
VBMGLVNBADCRDN-UHFFFAOYSA-N
1,3,5-tris(3,5-dibromophenyl)benzene
591077
Crystal-Powder at 20°C

Chemical Identifiers

29102-67-8
779.784
VBMGLVNBADCRDN-UHFFFAOYSA-N
1,3,5-tris(3,5-dibromophenyl)benzene
C24H12Br6
MFCD13193235
591077
C1=C(C=C(C=C1C2=CC(=CC(=C2)Br)Br)C3=CC(=CC(=C3)Br)Br)C4=CC(=CC(=C4)Br)Br
Safety and Handling

Safety and Handling

TSCA : No

Documents