missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Trimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Maybridge

$109.89 - $109.89

Chemical Identifiers

CAS 59340-27-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H9ClN2O2S
Formula Weight 208.67
MDL Number MFCD02180944
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-sulfonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC14703DE Fisher Scientific Company
CC14703DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC14703DA SDS Fisher Scientific Company
CC14703DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC14703CB SDS Fisher Scientific Company
CC14703CB
Amber Glass Bottle 250mg Each for $109.89
Specifications Description & Specifications

Chemical Identifiers

59340-27-1
100.0
C6H9ClN2O2S
MFCD02180944
97%
96.5
97%
208.67
1, 3, 5-Trimethyl-1H-pyrazole-4-sulfonyl chloride

Specifications

208.67
100.0
MFCD02180944
Amber Glass Bottle
59340-27-1
97%
5g
97%
96.5
C6H9ClN2O2S
1, 3, 5-Trimethyl-1H-pyrazole-4-sulfonyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger