Please login to your online account to display your discounted pricing

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC14701DA

 View more versions of this product

Catalog No. CC14701DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 154.17
Quantity 1g
Assay 97%
CAS 1125-29-7
Assay Percent Range 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Health Hazard 1 Warning
Molecular Formula C7H10N2O2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O2
Formula Weight 154.17
Accessories

Warning

SDS & Safety Documents