Please login to your online account to display your discounted pricing

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC14701CB

 View more versions of this product

Catalog No. CC14701CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.17
Quantity 250mg
Assay 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C7H10N2O2
CAS 1125-29-7
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O2
Formula Weight 154.17
Accessories

Warning