missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC14704DE
Specifications
1, 3, 5-Trimethyl-1H-pyrazole-4-carbaldehyde | |
97% | |
C7H10N2O | |
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h | |
CC1=C(C(=NN1C)C)C=O | |
138.17 | |
138.17 |
2644-93-1 | |
Amber Glass Bottle | |
5g | |
HMTUBXVXHHITGO-UHFFFAOYSA-N | |
1,3,5-trimethylpyrazole-4-carbaldehyde | |
736515 | |
97% |
Chemical Identifiers
2644-93-1 | |
138.17 | |
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h | |
1,3,5-trimethylpyrazole-4-carbaldehyde |
C7H10N2O | |
HMTUBXVXHHITGO-UHFFFAOYSA-N | |
736515 | |
CC1=C(C(=NN1C)C)C=O |