missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine, 97%, Maybridge™

$191.45 - $191.45

Chemical Identifiers

CAS 352018-93-0
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD02055811
InChI Key DGVBHLRORWEBLP-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1,3,5-trimethyl-1h-pyrazol-4-yl methylamine, 1,3,5-trimethylpyrazol-4-yl methanamine, 1,3,5-trimethylpyrazol-4-yl methylamine, 1-1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1-1,3,5-trimethylpyrazol-4-yl methanamine, 1h-pyrazole-4-methanamine, 1,3,5-trimethyl, c-1,3,5-trimethyl-1h-pyrazol-4-yl-methylamine, trimethyl-1h-pyrazol-4-yl methanamine, trimethylpyrazol-4-yl methanamine
PubChem CID 2776430
IUPAC Name 1-(1,3,5-trimethyl-1H-pyrazol-4-yl)methanamine
SMILES CN1N=C(C)C(CN)=C1C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14713CB
SDS View Documents Promotion Details
Fisher Scientific Company
CC14713CB
250mg Amber Glass Bottle
Each for $191.45
 
Description

Description

Specifications

Chemical Identifiers

352018-93-0
139.20
DGVBHLRORWEBLP-UHFFFAOYSA-N
2776430
CN1N=C(C)C(CN)=C1C
C7H13N3
MFCD02055811
1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1,3,5-trimethyl-1h-pyrazol-4-yl methylamine, 1,3,5-trimethylpyrazol-4-yl methanamine, 1,3,5-trimethylpyrazol-4-yl methylamine, 1-1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1-1,3,5-trimethylpyrazol-4-yl methanamine, 1h-pyrazole-4-methanamine, 1,3,5-trimethyl, c-1,3,5-trimethyl-1h-pyrazol-4-yl-methylamine, trimethyl-1h-pyrazol-4-yl methanamine, trimethylpyrazol-4-yl methanamine
1-(1,3,5-trimethyl-1H-pyrazol-4-yl)methanamine

Specifications

Amber Glass Bottle
352018-93-0
100.0
C7H13N3
1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1,3,5-trimethyl-1h-pyrazol-4-yl methylamine, 1,3,5-trimethylpyrazol-4-yl methanamine, 1,3,5-trimethylpyrazol-4-yl methylamine, 1-1,3,5-trimethyl-1h-pyrazol-4-yl methanamine, 1-1,3,5-trimethylpyrazol-4-yl methanamine, 1h-pyrazole-4-methanamine, 1,3,5-trimethyl, c-1,3,5-trimethyl-1h-pyrazol-4-yl-methylamine, trimethyl-1h-pyrazol-4-yl methanamine, trimethylpyrazol-4-yl methanamine
CN1N=C(C)C(CN)=C1C
139.20
139.2
(1, 3, 5-Trimethyl-1H-pyrazol-4-yl)methylamine
250mg
96.5
97%
MFCD02055811
DGVBHLRORWEBLP-UHFFFAOYSA-N
1-(1,3,5-trimethyl-1H-pyrazol-4-yl)methanamine
2776430
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications