Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trimethyl-1,4-cyclohexadiene, 98%

Chemical Identifiers

Chemical Name or Material 1,3,5-Trimethyl-1,4-cyclohexadiene
CAS 4074-23-1
Molecular Formula C9H14
Formula Weight 122.21
MDL Number MFCD04974021
UN Number UN3295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2592106

  SDS
alfa aesar™
H2592106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2592114

  SDS
alfa aesar™
H2592114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.21
Chemical Name or Material 1,3,5-Trimethyl-1,4-cyclohexadiene
Assay 98%
Molecular Formula C9H14
MDL Number MFCD04974021
CAS 4074-23-1
UN Number UN3295
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Flash Point 35°C (95°F)
Boiling Point 172°C
Quantity 5g

Applications
1,3,5-Trimethyl-1,4-cyclohexadiene is used as intermediates in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Warning