missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Trimethoxybenzene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 160552500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 3, 5-Trimethoxybenzene | |
98.5 | |
85g/mL | |
85°C | |
98.5% min. (GC) | |
C9H12O3 | |
MFCD00008385 | |
06, 1101 | |
Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents,soluble in methanol | |
COC1=CC(OC)=CC(OC)=C1 | |
168.19 | |
CHEBI:31038 | |
99% |
621-23-8 | |
100.0 | |
255.0°C | |
Authentic | |
Glass bottle | |
C6H3(OCH3)3 | |
250g | |
phloroglucinol trimethyl ether, sym-trimethoxybenzene, benzene, 1,3,5-trimethoxy, 1,3,5-trimethyoxybenzene, unii-00vji3vg3d, 1,3,5-trimethoxy benzene, o,o,o-1,3,5-trimethylresorcinol, 00vji3vg3d, dsstox_cid_25963, dsstox_rid_81259 | |
LKUDPHPHKOZXCD-UHFFFAOYSA-N | |
1,3,5-trimethoxybenzene | |
69301 | |
168.19 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 210-673-4
RUO â Research Use Only