Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trifluoro-2-nitrobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1,3,5-Trifluoro-2-nitrobenzene
CAS 315-14-0
Molecular Formula C6H2F3NO2
Formula Weight 177.08
MDL Number MFCD00014687
UN Number UN2810
Beilstein 2106869
Synonym 2-Nitro-1,3,5-trifluorobenzene; 2,4,6-Trifluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1703506

  SDS
alfa aesar™
A1703506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1703514

  SDS
alfa aesar™
A1703514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1703503

  SDS
alfa aesar™
A1703503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1,3,5-Trifluoro-2-nitrobenzene
Flash Point 77°C (170°F)
Boiling Point 171° to 172°C
Melting Point 2° to 5°C
Quantity 5g
CAS 315-14-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Assay 98%
Molecular Formula C6H2F3NO2
Formula Weight 177.08
MDL Number MFCD00014687
UN Number UN2810
Beilstein 2106869
Synonym 2-Nitro-1,3,5-trifluorobenzene; 2,4,6-Trifluoronitrobenzene
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.