missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Triethynylbenzene, TCI America™

Manufacturer:  TCI America T27601G

 View more versions of this product

Catalog No. T27601G



Specifications

Specifications

1,3,5-Triethynylbenzene
105°C
7567-63-7
MFCD00068621
ZDRMMTYSQSIGRY-UHFFFAOYSA-N
1,3,5-triethynylbenzene
139048
Crystal-Powder at 20°C
Undesignated
1g
C12H6
benzene, 1,3,5-triethynyl, benzene,1,3,5-triethynyl, 1,3,5-phenylene-triacetylene, acmc-209ozh, 1,3,5-triethynyl-benzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene 1g, benzene, 1,3,5-triethynyl-8ci, 9ci
C#CC1=CC(=CC(=C1)C#C)C#C
150.18
150.18
≥98.0% (GC)

Chemical Identifiers

7567-63-7
150.18
ZDRMMTYSQSIGRY-UHFFFAOYSA-N
139048
C#CC1=CC(=CC(=C1)C#C)C#C
C12H6
MFCD00068621
benzene, 1,3,5-triethynyl, benzene,1,3,5-triethynyl, 1,3,5-phenylene-triacetylene, acmc-209ozh, 1,3,5-triethynyl-benzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene 1g, benzene, 1,3,5-triethynyl-8ci, 9ci
1,3,5-triethynylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents