missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Triethylbenzene, TCI America™

Manufacturer:  TCI America T235225ML

 View more versions of this product

Catalog No. T235225ML



Specifications

Specifications

1,3,5-Triethylbenzene
Colorless
25mL
C12H18
3082
WJYMPXJVHNDZHD-UHFFFAOYSA-N
1,3,5-triethylbenzene
7602
Clear Liquid at 20°C
215°C
-66°C
102-25-0
MFCD00009261
benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
CCC1=CC(=CC(=C1)CC)CC
162.276
162.28
≥90.0% (GC)

Chemical Identifiers

102-25-0
162.276
WJYMPXJVHNDZHD-UHFFFAOYSA-N
7602
CCC1=CC(=CC(=C1)CC)CC
C12H18
MFCD00009261
benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
1,3,5-triethylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3427

TSCA : Yes

Documents