missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Triethylbenzene, 95%

$76.65 - $334.64

Chemical Identifiers

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(=CC(=C1)CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6087506 View Documents Alfa Aesar™
H6087506
5g Each for $76.65
 
AAH6087514 View Documents Alfa Aesar™
H6087514
25g Each for $334.64
 
Description

Description

Applications
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

Solubility
Insoluble in water. Soluble in alcohol, ether.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Chemical Identifiers

102-25-0
162.276
WJYMPXJVHNDZHD-UHFFFAOYSA-N
7602
CCC1=CC(=CC(=C1)CC)CC
C12H18
MFCD00009261
benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
1,3,5-triethylbenzene

Specifications

76°C (169°F)
5g
C12H18
UN3082
benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
CCC1=CC(=CC(=C1)CC)CC
162.276
162.27
1,3,5-Triethylbenzene
215°C
102-25-0
MFCD00009261
2039481
WJYMPXJVHNDZHD-UHFFFAOYSA-N
1,3,5-triethylbenzene
7602
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA