Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trichlorobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1,3,5-Trichlorobenzene
CAS 108-70-3
Molecular Formula C6H3Cl3
Formula Weight 181.45g/mol
MDL Number MFCD00000585
UN Number UN3077
Beilstein 1635233
Merck Index 14,9632
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0213218

 
alfa aesar™
L02132-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0213230

 
alfa aesar™
L02132-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 208°C
Melting Point 62° to 65°C
Flash Point 126°C (258°F)
Quantity 50g
Chemical Name or Material 1,3,5-Trichlorobenzene
CAS 108-70-3
Assay 98%
Molecular Formula C6H3Cl3
Formula Weight 181.45g/mol
MDL Number MFCD00000585
UN Number UN3077
Beilstein 1635233
Merck Index 14,9632
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number DC2100100
TSCA TSCA

Applications
1,3,5-Trichlorobenzene is used in metal treatments, in industrial deodorants, in cleaners for drains.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.