Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trichloro-2-iodobenzene, 96%

$43.84 - $141.64

Chemical Identifiers

Chemical Name or Material 1,3,5-Trichloro-2-iodobenzene
CAS 6324-50-1
Molecular Formula C6H2Cl3I
Formula Weight 307.35
MDL Number MFCD00052857
Beilstein 1868441
Synonym 1-Iodo-2,4,6-trichlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1197503

  SDS
alfa aesar™
L1197503
1g Each for $43.84

AAL1197506

  SDS
alfa aesar™
L1197506
5g Each for $141.64
Description & Specifications

Specifications

Formula Weight 307.35
MDL Number MFCD00052857
Molecular Formula C6H2Cl3I
Chemical Name or Material 1,3,5-Trichloro-2-iodobenzene
Assay 96%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Iodo-2,4,6-trichlorobenzene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Beilstein 1868441
Melting Point 52° to 57°C
Health Hazard 1 Warning
CAS 6324-50-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.