missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Trichloro-2-iodobenzene, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1197506

 View more versions of this product

Catalog No. AAL1197506



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,3,5-Trichloro-2-iodobenzene
52°C to 57°C
MFCD00052857
1868441
2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene
C1=C(C=C(C(=C1Cl)I)Cl)Cl
307.336
307.35
6324-50-1
C6H2Cl3I
5 g
Light sensitive
XXFFSBRDPQFIPO-UHFFFAOYSA-N
1,3,5-trichloro-2-iodobenzene
232054
96%

Chemical Identifiers

6324-50-1
307.336
XXFFSBRDPQFIPO-UHFFFAOYSA-N
232054
C1=C(C=C(C(=C1Cl)I)Cl)Cl
C6H2Cl3I
MFCD00052857
2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene
1,3,5-trichloro-2-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only