missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trichloro-2-iodobenzene, 96%

Manufacturer:  Alfa Aesar™ L1197503

 View more versions of this product

Catalog No. AAL1197503



Specifications

Specifications

1,3,5-Trichloro-2-iodobenzene
1g
C6H2Cl3I
1868441
XXFFSBRDPQFIPO-UHFFFAOYSA-N
1,3,5-trichloro-2-iodobenzene
232054
96%
52°C to 57°C
6324-50-1
MFCD00052857
2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene
C1=C(C=C(C(=C1Cl)I)Cl)Cl
307.336
307.35

Chemical Identifiers

6324-50-1
307.336
XXFFSBRDPQFIPO-UHFFFAOYSA-N
232054
C1=C(C=C(C(=C1Cl)I)Cl)Cl
C6H2Cl3I
MFCD00052857
2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene
1,3,5-trichloro-2-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents