Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Trichloro-2,4,6-trifluorobenzene, 98%

$57.09 - $795.81

Chemical Identifiers

Chemical Name or Material 1,3,5-Trichloro-2,4,6-trifluorobenzene
CAS 319-88-0
Molecular Formula C6Cl3F3
Formula Weight 235.42
MDL Number MFCD00000587
Beilstein 958307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1686414

 
alfa aesar™
A1686414
25g Each for $233.97

AAA1686406

 
alfa aesar™
A1686406
5g Each for $57.09

AAA1686422

 
alfa aesar™
A1686422
100g Each for $795.81
Description & Specifications

Specifications

Formula Weight 235.42
Chemical Name or Material 1,3,5-Trichloro-2,4,6-trifluorobenzene
Assay 98%
Molecular Formula C6Cl3F3
MDL Number MFCD00000587
CAS 319-88-0
Beilstein 958307
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 79° to 80°C (12mmHg)
Melting Point 62° to 64°C
Flash Point 102°C (215°F)
RTECS Number DC2450000
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.