Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Tribenzoylbenzene, 96%

$241.08 - $241.08

Chemical Identifiers

Chemical Name or Material 1,3,5-Tribenzoylbenzene
CAS 25871-69-6
Molecular Formula C27H18O3
Formula Weight 390.44
MDL Number MFCD00016577
Beilstein 1891480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1994106

  SDS
alfa aesar™
A1994106
5g Each for $241.08
Description & Specifications

Specifications

Formula Weight 390.44
Melting Point 114° to 121°C
Assay 96%
Chemical Name or Material 1,3,5-Tribenzoylbenzene
Molecular Formula C27H18O3
CAS 25871-69-6
Beilstein 1891480
MDL Number MFCD00016577
Quantity 5g

Applications
It is used as a ligand in the synthesis and characterization of three new coordination polymers from the tritopic ligand.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.