Please login to your online account to display your discounted pricing

1,3,5-Triazine-2,4,6-trithiol trisodium salt, approx. 15% solution in water, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426540010

 View more versions of this product

Catalog No. AC426540010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 243.22
Quantity 1L
Color Colorless to light yellow
Density 1.1200g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C3N3Na3S3
CAS 17766-26-6
Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt

Specifications

Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt
CAS 17766-26-6
Molecular Formula C3N3Na3S3
Formula Weight 243.22