Please login to your online account to display your discounted pricing

1,3,5-Triazine-2,4,6-trithiol trisodium salt, approx. 15% solution in water, ACROS Organics™

$64.82 - $64.82

Chemical Identifiers

Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt
CAS 17766-26-6
Molecular Formula C3N3Na3S3
Formula Weight 243.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426540025

  SDS
thermo fisher scientific
AC426540025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426540010

  SDS
thermo fisher scientific
AC426540010
Glass bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426542500

  SDS
thermo fisher scientific
AC426542500
Glass bottle 250mL Each for $64.82
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 243.22
Quantity 2.5L
Color Colorless to light yellow
Density 1.1200g/mL
Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt
CAS 17766-26-6
Molecular Formula C3N3Na3S3
Infrared Spectrum Authentic
Packaging Glass bottle