Please login to your online account to display your discounted pricing

1,3,5-Triazine-2,4,6-trithiol trisodium salt, approx. 15% solution in water, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426542500

 View more versions of this product

Catalog No. AC426542500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 243.22
Quantity 250mL
Color Colorless to light yellow
Density 1.1200g/mL
CAS 17766-26-6
Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C3N3Na3S3

Specifications

Chemical Name or Material 1,3,5-Triazine-2,4,6-trithiol trisodium salt
CAS 17766-26-6
Molecular Formula C3N3Na3S3
Formula Weight 243.22