missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Triacetylbenzene, TCI America™

Manufacturer:  TCI America T133725G

 View more versions of this product

Catalog No. T133725G



Specifications

Specifications

1,3,5-Triacetylbenzene
163°C
779-90-8
MFCD00008741
HSOAIPRTHLEQFI-UHFFFAOYSA-N
1-(3,5-diacetylphenyl)ethanone
69904
Crystal-Powder at 20°C
Yellow
25g
C12H12O3
1,3,5-triacetylbenzene, 1,1',1-benzene-1,3,5-triyl triethanone, 1,3,5,triacetylbenzene, benzene, 1,3,5-triacetyl, 1-3,5-diacetylphenyl ethanone, 1,5-triacetylbenzene, benzene,3,5-triacetyl, acmc-1bc4n, cambridge id 6741393, ethanone, 1,1',1-1,3,5-benzenetriyl tris
CC(=O)C1=CC(=CC(=C1)C(=O)C)C(=O)C
204.225
204.23
≥98.0% (GC)

Chemical Identifiers

779-90-8
204.225
HSOAIPRTHLEQFI-UHFFFAOYSA-N
69904
CC(=O)C1=CC(=CC(=C1)C(=O)C)C(=O)C
C12H12O3
MFCD00008741
1,3,5-triacetylbenzene, 1,1',1-benzene-1,3,5-triyl triethanone, 1,3,5,triacetylbenzene, benzene, 1,3,5-triacetyl, 1-3,5-diacetylphenyl ethanone, 1,5-triacetylbenzene, benzene,3,5-triacetyl, acmc-1bc4n, cambridge id 6741393, ethanone, 1,1',1-1,3,5-benzenetriyl tris
1-(3,5-diacetylphenyl)ethanone
Safety and Handling

Safety and Handling

TSCA : No

Documents