missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Tri-tert-butylbenzene 98.0+%, TCI America™
Supplier: TCI America T163325G
Specifications
1,3,5-Tri-tert-butylbenzene | |
74°C | |
248°C | |
MFCD00008831 | |
1,3,5-tri-tert-butylbenzene, benzene, 1,3,5-tris 1,1-dimethylethyl, 1,3,5-tri-t-butylbenzene, benzene, 1,3,5-tri-tert-butyl, 1,3,5-tris tert-butyl benzene, pubchem13719, intermediates-zcf02115, acmc-209cw6, 2,4,6-tri-tert-butylphenyl, ksc490m7l | |
CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C | |
246.438 | |
246.44 | |
Crystalline Powder |
1460-02-2 | |
White-Yellow | |
C18H30 | |
25 g | |
GUFMBISUSZUUCB-UHFFFAOYSA-N | |
1,3,5-tritert-butylbenzene | |
15089 | |
≥98.0% (GC) |
Chemical Identifiers
1460-02-2 | |
246.438 | |
GUFMBISUSZUUCB-UHFFFAOYSA-N | |
15089 | |
CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C |
C18H30 | |
MFCD00008831 | |
1,3,5-tri-tert-butylbenzene, benzene, 1,3,5-tris 1,1-dimethylethyl, 1,3,5-tri-t-butylbenzene, benzene, 1,3,5-tri-tert-butyl, 1,3,5-tris tert-butyl benzene, pubchem13719, intermediates-zcf02115, acmc-209cw6, 2,4,6-tri-tert-butylphenyl, ksc490m7l | |
1,3,5-tritert-butylbenzene |
Safety and Handling
EINECSNumber : (3)-3427
TSCA : No