missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6407803

 View more versions of this product

Catalog No. AAH6407803


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine
Colorless
94°C (0.7 mmHg)
1.466
1 g
1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine
CCCN1CN(CCC)CN(CCC)C1
213.37
213.37
Liquid
13036-81-2
0.880 g/mL
C12H27N3
MFCD00040461
111423
DPHBVWJGMBYPMK-UHFFFAOYSA-N
1,3,5-tripropyl-1,3,5-triazinane
83068
96%

Chemical Identifiers

13036-81-2
213.37
DPHBVWJGMBYPMK-UHFFFAOYSA-N
83068
CCCN1CN(CCC)CN(CCC)C1
C12H27N3
MFCD00040461
1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine
1,3,5-tripropyl-1,3,5-triazinane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 235-904-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions