missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5-Tri-n-propylhexahydro-1,3,5-triazine, 96%

Manufacturer:  Alfa Aesar™ H6407806

 View more versions of this product

Catalog No. AAH6407806


 

Specifications

Specifications

1,3,5-Tri-n-propylhexahydro-1,3,5-triazine
94°C (0.7mmHg)
5g
1.4660
C12H27N3
111423
DPHBVWJGMBYPMK-UHFFFAOYSA-N
1,3,5-tripropyl-1,3,5-triazinane
83068
Liquid
0.880g/mL
Colorless
13036-81-2
96%
MFCD00040461
Hexahydro-1,3,5-tri-n-propyl-1,3,5-triazine; 1,3,5-Tri-n-propyl-1,3,5-triazinane
CCCN1CN(CN(C1)CCC)CCC
213.369
213.37

Chemical Identifiers

13036-81-2
213.369
DPHBVWJGMBYPMK-UHFFFAOYSA-N
83068
C12H27N3
MFCD00040461
Hexahydro-1,3,5-tri-n-propyl-1,3,5-triazine; 1,3,5-Tri-n-propyl-1,3,5-triazinane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents