Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole, 97%

$37.26 - $441.77

Chemical Identifiers

Chemical Name or Material 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole
CAS 175205-50-2
Molecular Formula C12H11Cl2N
Formula Weight 240.13
MDL Number MFCD00084999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2512503

  SDS
alfa aesar™
B2512503
1g Each for $37.26

AAB2512514

  SDS
alfa aesar™
B2512514
25g Each for $441.77

AAB2512506

  SDS
alfa aesar™
B2512506
5g Each for $126.68
Description & Specifications

Specifications

Formula Weight 240.13
Chemical Name or Material 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole
Assay 97%
Molecular Formula C12H11Cl2N
MDL Number MFCD00084999
CAS 175205-50-2
Melting Point 78° to 80°C
Quantity 1g