Please login to your online account to display your discounted pricing

1-(3,5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD08850DA

 View more versions of this product

Catalog No. CD08850DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.09
Quantity 1g
Assay 97%
CAS Max % 100.0
CAS 175136-79-5
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde
CAS Min % 96.5
MDL Number MFCD00174281
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H7Cl2NO

Specifications

Chemical Name or Material 1-(3, 5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde
CAS 175136-79-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7Cl2NO
Formula Weight 240.09
MDL Number MFCD00174281