Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole, 97%

$51.08 - $170.27

Chemical Identifiers

Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
CAS 175136-60-4
Molecular Formula C12H7F6N
Formula Weight 279.19
MDL Number MFCD00053048
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1575703

  SDS
alfa aesar™
A1575703
1g Each for $51.08

AAA1575706

  SDS
alfa aesar™
A1575706
5g Each for $170.27
Description & Specifications

Specifications

Formula Weight 279.19
Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
Assay 97%
Molecular Formula C12H7F6N
MDL Number MFCD00053048
CAS 175136-60-4
Melting Point 41° to 43°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.