missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1575706

 View more versions of this product

Catalog No. AAA1575706



Specifications

Specifications

1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
5g
C12H7F6N
1-3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, pubchem9334, maybridge1_002820, acmc-1c4qv, 3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-di trifluoromethyl phenyl-1h-pyrrole
C1=CN(C=C1)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
279.185
279.19
41°C to 43°C
175136-60-4
MFCD00053048
BKSCRBOUGNMEKQ-UHFFFAOYSA-N
1-[3,5-bis(trifluoromethyl)phenyl]pyrrole
2736181
97%

Chemical Identifiers

175136-60-4
279.185
BKSCRBOUGNMEKQ-UHFFFAOYSA-N
2736181
C1=CN(C=C1)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
C12H7F6N
MFCD00053048
1-3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, pubchem9334, maybridge1_002820, acmc-1c4qv, 3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-di trifluoromethyl phenyl-1h-pyrrole
1-[3,5-bis(trifluoromethyl)phenyl]pyrrole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents