missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6258303

 View more versions of this product

Catalog No. AAH6258303



Specifications

Specifications

1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea
1g
MFCD04069619
Solid
White
978-65-4
366.23
97%

Chemical Identifiers

978-65-4
MFCD04069619
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents