missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea, 97%, Thermo Scientific™

$139.34 - $139.34

Chemical Identifiers

CAS 978-65-4
MDL Number MFCD04069619
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62583MD
View Documents Promotion Details
Thermo Scientific Chemicals
H62583MD
250 mg
Each for $139.34
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

978-65-4
MFCD04069619

Specifications

978-65-4
MFCD04069619
366.23
Solid
White
250 mg
97%
1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'