missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H6258306

 View more versions of this product

Catalog No. AAH6258306



Specifications

Specifications

1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea
5g
MFCD04069619
Solid
White
978-65-4
366.23
97%

Chemical Identifiers

978-65-4
MFCD04069619
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents