Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole, 97%

$143.30 - $472.55

Chemical Identifiers

Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
CAS 175205-51-3
Molecular Formula C12H11F6N
Formula Weight 283.22
MDL Number MFCD00084902
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2508706

 
alfa aesar™
B2508706
5g Each for $143.30

AAB2508714

 
alfa aesar™
B2508714
25g Each for $472.55
Description & Specifications

Specifications

Formula Weight 283.22
Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Assay 97%
Molecular Formula C12H11F6N
MDL Number MFCD00084902
CAS 175205-51-3
Boiling Point 78° to 80°C (0.1mmHg)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.