Please login to your online account to display your discounted pricing

1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC209350010

 View more versions of this product

Catalog No. AC209350010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid or Low Melting Solid
Formula Weight 330.33
Quantity 1g
Color Light yellow
Density 1.16g/mL
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Synonym Triallyl 1, 3, 5-benzenetricarboxylate
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
Molecular Formula C18H18O6
CAS 17832-16-5
Linear Formula C6H3(CO2CH2CH=CH2)3
Assay Percent Range 98%
MDL Number MFCD00008647
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
CAS 17832-16-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C18H18O6
Linear Formula C6H3(CO2CH2CH=CH2)3
Formula Weight 330.33
MDL Number MFCD00008647
Synonym Triallyl 1, 3, 5-benzenetricarboxylate