Please login to your online account to display your discounted pricing

1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
CAS 17832-16-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C18H18O6
Linear Formula C6H3(CO2CH2CH=CH2)3
Formula Weight 330.33
MDL Number MFCD00008647
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209350010

  SDS
thermo fisher scientific
AC209350010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC209350050

  SDS
thermo fisher scientific
AC209350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid or Low Melting Solid
Assay ≥97.5%
Formula Weight 330.33
Quantity 1g
Color Light yellow
Density 1.16g/mL
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C18H18O6
Linear Formula C6H3(CO2CH2CH=CH2)3
MDL Number MFCD00008647
CAS 17832-16-5
Synonym Triallyl 1, 3, 5-benzenetricarboxylate