Please login to your online account to display your discounted pricing

1,3,5-Benzenetricarboxylic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: fisher scientific acros  105350050

 View more versions of this product

Catalog No. AC105350050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 210.14
Quantity 5g
Beilstein 09, 978
Packaging Glass Bottle
CAS Max % 100.0
Linear Formula C6H3(CO2H)3
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid
Assay Percent Range 98%
Molecular Formula C9H6O6
MDL Number MFCD00002517
CAS 554-95-0
Synonym Trimesic acid
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid
CAS 554-95-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H6O6
Linear Formula C6H3(CO2H)3
Formula Weight 210.14
MDL Number MFCD00002517
Synonym Trimesic acid
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents