missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0725522

 View more versions of this product

Catalog No. AAL0725522


Description

Description

1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.
Specifications

Specifications

1,3,5-Benzenetricarbonyl chloride
34°C to 38°C
>110°C (230°F)
MFCD00000679
UN3261
Moisture sensitive
Soluble in chloroform(soluble10%,clear to very slightly hazy,colorless to faintly yellow) and toluene. Solubility in water: Reacts.
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
265.47
265.48
4422-95-1
180°C (16 mmHg)
C9H3Cl3O3
100 g
2940936
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
UWCPYKQBIPYOLX-UHFFFAOYSA-N
benzene-1,3,5-tricarbonyl chloride
78138
≥98%

Chemical Identifiers

4422-95-1
265.47
UWCPYKQBIPYOLX-UHFFFAOYSA-N
78138
C9H3Cl3O3
MFCD00000679
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 224-594-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only