missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5,7-Cyclooctatetraene, 98%, stab. with 0.1% Hydroquinone, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2263614

 View more versions of this product

Catalog No. AAB2263614



Description

Description

1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

Solubility
Not miscible or difficult to mix in water.

Notes
Air & light sensitive. Store at --20°C. Store away from oxidizing agent, air, heat, ultra violet radiation, free radical intiators.
Specifications

Specifications

1,3,5,7-Cyclooctatetraene
629-20-9
0.925
20°C (68°F)
C8H8
MFCD00004161
UN2358
Air and light sensitive
Not miscible or difficult to mix in water.
C1=C/C=C\C=C/C=C\1
104.15
104.15
stabilized with 0.1% Hydroquinone
-5°C
142°C to 143°C
Unpleasant
1.537
25 g
2496800
1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
KDUIUFJBNGTBMD-DLMDZQPMSA-N
cyclooctatetraene
637866
98%

Chemical Identifiers

629-20-9
104.15
KDUIUFJBNGTBMD-DLMDZQPMSA-N
637866
C1=C/C=C\C=C/C=C\1
C8H8
MFCD00004161
1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
cyclooctatetraene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOOCTATETRAENE

EINECSNumber : 211-080-3

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only