Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3,5,5-Tetramethyl-1,3-cyclohexadiene, 94%

Chemical Identifiers

Chemical Name or Material 1,3,5,5-Tetramethyl-1,3-cyclohexadiene
CAS 4724-89-4
Molecular Formula C10H16
Formula Weight 136.24
MDL Number MFCD00045518
UN Number UN3295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2161703

  SDS
alfa aesar™
B2161703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2161706

  SDS
alfa aesar™
B2161706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.24
Flash Point 52°C (126°F)
Quantity 1g
CAS 4724-89-4
UN Number UN3295
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H16
Chemical Name or Material 1,3,5,5-Tetramethyl-1,3-cyclohexadiene
TSCA TSCA
MDL Number MFCD00045518
Assay 94%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.