Please login to your online account to display your discounted pricing

1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine, 97%, Maybridge

$108.46 - $108.46

Chemical Identifiers

Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H19N3
Formula Weight 205.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC35913DE

 
fisher scientific company
CC35913DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC35913CB

 
fisher scientific company
CC35913CB
Amber Glass Bottle 250mg Each for $108.46

CC35913DA

 
fisher scientific company
CC35913DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.3
Quantity 5g
Assay 97%
Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Packaging Amber Glass Bottle
Molecular Formula C12H19N3
CAS 672325-37-0
Assay Percent Range 97%