Please login to your online account to display your discounted pricing

1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC35913DA

 View more versions of this product

Catalog No. CC35913DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.3
Quantity 1g
Assay 97%
CAS 672325-37-0
Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Assay Percent Range 97%
Molecular Formula C12H19N3
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H19N3
Formula Weight 205.3