Please login to your online account to display your discounted pricing

1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC35913CB

 View more versions of this product

Catalog No. CC35913CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 205.3
Quantity 250mg
Assay 97%
Assay Percent Range 97%
CAS 672325-37-0
Packaging Amber Glass Bottle
Molecular Formula C12H19N3
Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine

Specifications

Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H19N3
Formula Weight 205.3