Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3,4-Methylenedioxy)phenyl-2-butanone, 96%

$42.50 - $121.74

Chemical Identifiers

Chemical Name or Material 1-(3,4-Methylenedioxy)phenyl-2-butanone
CAS 23023-13-4
Molecular Formula C11H12O3
Formula Weight 192.22
MDL Number MFCD02093417
Beilstein 6508240
Synonym 1-(1,3-Benzodioxol-5-yl)-2-butanone; Ethyl piperonyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1364303

  SDS
alfa aesar™
L1364303
1g Each for $42.50

AAL1364306

  SDS
alfa aesar™
L1364306
5g Each for $121.74
Description & Specifications

Specifications

Chemical Name or Material 1-(3,4-Methylenedioxy)phenyl-2-butanone
CAS 23023-13-4
TSCA TSCA
Assay 96%
Molecular Formula C11H12O3
Formula Weight 192.22
MDL Number MFCD02093417
Beilstein 6508240
Synonym 1-(1,3-Benzodioxol-5-yl)-2-butanone; Ethyl piperonyl ketone
Boiling Point 98°C (0.1mmHg)
Quantity 1g

Applications
1-(3,4-Methylenedioxy)phenyl-2-butanone, is used as a pharmaceutical intermediate.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.