missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3,4-Methylenedioxy)phenyl-2-butanone, 96%

Manufacturer:  Alfa Aesar™ L1364303

 View more versions of this product

Catalog No. AAL1364303



Description

Description

Applications
1-(3,4-Methylenedioxy)phenyl-2-butanone, is used as a pharmaceutical intermediate.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

1-(3,4-Methylenedioxy)phenyl-2-butanone
1g
C11H12O3
6508240
SWKXHWCUDGJLNA-UHFFFAOYSA-N
1-(1,3-benzodioxol-5-yl)butan-2-one
89964
96%
98°C (0.1mmHg)
23023-13-4
MFCD02093417
2-butanone, 1-1,3-benzodioxol-5-yl, 1-1,3-benzodioxol-5-yl butan-2-one, 1-3,4-methylenedioxyphenyl-2-butanone, 1-1,3-benzodioxol-5-yl-2-butanone, 1-benzo d 1,3 dioxol-5-yl butan-2-one, 2-butanone,1-1,3-benzodioxol-5-yl, 1-2h-1,3-benzodioxol-5-yl butan-2-one, 1-3,4-methylenedioxyphenyl butanone-2, acmc-1cr8m, 3,4-methylendioxyphenylbutan-2-one
CCC(=O)CC1=CC2=C(C=C1)OCO2
192.214
192.22

Chemical Identifiers

23023-13-4
192.214
SWKXHWCUDGJLNA-UHFFFAOYSA-N
89964
CCC(=O)CC1=CC2=C(C=C1)OCO2
C11H12O3
MFCD02093417
2-butanone, 1-1,3-benzodioxol-5-yl, 1-1,3-benzodioxol-5-yl butan-2-one, 1-3,4-methylenedioxyphenyl-2-butanone, 1-1,3-benzodioxol-5-yl-2-butanone, 1-benzo d 1,3 dioxol-5-yl butan-2-one, 2-butanone,1-1,3-benzodioxol-5-yl, 1-2h-1,3-benzodioxol-5-yl butan-2-one, 1-3,4-methylenedioxyphenyl butanone-2, acmc-1cr8m, 3,4-methylendioxyphenylbutan-2-one
1-(1,3-benzodioxol-5-yl)butan-2-one
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents