Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3,4-Dichlorophenyl)piperazine, 98%

Chemical Identifiers

Chemical Name or Material 1-(3,4-Dichlorophenyl)piperazine
CAS 57260-67-0
Molecular Formula C10H12Cl2N2
Formula Weight 231.13
MDL Number MFCD00023128
UN Number UN3259
Beilstein 520169
Synonym 1,2-Dichloro-4-(1-piperazino)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0554406

 
alfa aesar™
L0554406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0554414

 
alfa aesar™
L0554414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.13
Chemical Name or Material 1-(3,4-Dichlorophenyl)piperazine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 98%
MDL Number MFCD00023128
CAS 57260-67-0
UN Number UN3259
Boiling Point 202°C (15mmHg)
Beilstein 520169
Synonym 1,2-Dichloro-4-(1-piperazino)benzene
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Quantity 5g
Molecular Formula C10H12Cl2N2
Melting Point 61° to 65°C
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.