missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3,4-Dichlorobenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5568514

 View more versions of this product

Catalog No. AAH5568514



Specifications

Specifications

1-(3,4-Dichlorobenzyl)piperazine
>110°C (230°F)
Colorless to Yellow
55513-17-2
MFCD03407488
PNDSYXGJCWKNFG-UHFFFAOYSA-N
1-[(3,4-dichlorophenyl)methyl]piperazine
2758074
Liquid
1.225 g/mL
124°C to 128°C (0.02mmHg)
25g
C11H14Cl2N2
1-3,4-dichloro-benzyl-piperazine
C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl
245.147
245.15
97%

Chemical Identifiers

55513-17-2
245.147
PNDSYXGJCWKNFG-UHFFFAOYSA-N
2758074
C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl
C11H14Cl2N2
MFCD03407488
1-3,4-dichloro-benzyl-piperazine
1-[(3,4-dichlorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents