Please login to your online account to display your discounted pricing

1-[3-(4-chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
CAS 169814-48-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNO2
Formula Weight 235.67
MDL Number MFCD01313507
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB04957DA

  SDS
fisher scientific company
SPB04957DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB04957EA

  SDS
fisher scientific company
SPB04957EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 235.67
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H10ClNO2
MDL Number MFCD01313507
CAS 169814-48-6